  • Search Results
  • Perturbation of copper homeostasis is instrumental in early developmental arrest of intraerythrocytic Plasmodium falciparum.
Perturbation of copper homeostasis is instrumental in early developmental arrest of intraerythrocytic Plasmodium falciparum.
BMC microbiology (2014-06-26)
Hiroko Asahi, Mohammed Essa Marghany Tolba, Masanobu Tanabe, Sumio Sugano, Kazumi Abe, Fumihiko Kawamoto
ABSTRACT
Malaria continues to be a devastating disease. The elucidation of factors inducing asexual growth versus arrest of Plasmodium falciparum can provide information about the development of the parasite, and may help in the search for novel malaria medication. Based on information from genome-wide transcriptome profiling of different developmental stages of P. falciparum, we investigated the critical importance of copper homeostasis in the developmental succession of P. falciparum with regard to three aspects of copper function. These were:1) inhibition of copper-binding proteins, 2) copper-ion chelation, and 3) down-regulated expression of genes encoding copper-binding proteins associated with a specific growth-promoting factor. Inhibition of copper-binding proteins with tetrathiomolybdate (TTM) caused cessation of growth of the parasite. TTM arrested the parasite irreversibly during the trophozoite to schizont stage progression. Target molecules for TTM may be present in P. falciparum. The involvement of copper ions in developmental arrest was also investigated by copper-ion chelating methods, which indicated a critical function of reduced copper ions (Cu1+) in the parasite during the early developmental stage. Copper ions, not only in the parasite but also in host cells, were targets of the chelators. Chelation of Cu1+caused blockage of trophozoite progression from the ring stage. Profound growth arrest was detected in parasites cultured in a chemically defined medium containing hexadecanoic acid alone as a growth-promoting factor. This developmental arrest was associated with down-regulated expression of genes encoding copper-binding proteins. Cis-9-octadecenoic acid completely prevented the down-regulation of gene expression and developmental arrest that were observed with the use of hexadecanoic acid. The critical importance of copper homeostasis in early developmental stages of P. falciparum was confirmed. Perturbation of copper homeostasis induced profound and early developmental arrest of P. falciparum. These findings should help to elucidate the mechanisms behind the development of P. falciparum, and may be applied in the development of effective antimalarial strategies.
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sorbitol, European Pharmacopoeia (EP) Reference Standard
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Neocuproine, ≥98%
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
D-Sorbitol, ≥98%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-Sorbitol, BioXtra, ≥98%
Sigma-Aldrich
D-Sorbitol, for molecular biology, ≥98%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, 99%
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C