  • Search Results
  • Inhibition of the SR protein-phosphorylating CLK kinases of Plasmodium falciparum impairs blood stage replication and malaria transmission.
Inhibition of the SR protein-phosphorylating CLK kinases of Plasmodium falciparum impairs blood stage replication and malaria transmission.
PloS one (2014-09-05)
Selina Kern, Shruti Agarwal, Kilian Huber, André P Gehring, Benjamin Strödke, Christine C Wirth, Thomas Brügl, Liliane Onambele Abodo, Thomas Dandekar, Christian Doerig, Rainer Fischer, Andrew B Tobin, Mahmood M Alam, Franz Bracher, Gabriele Pradel
ABSTRACT
Cyclin-dependent kinase-like kinases (CLKs) are dual specificity protein kinases that phosphorylate Serine/Arginine-rich (SR) proteins involved in pre-mRNA processing. Four CLKs, termed PfCLK-1-4, can be identified in the human malaria parasite Plasmodium falciparum, which show homology with the yeast SR protein kinase Sky1p. The four PfCLKs are present in the nucleus and cytoplasm of the asexual blood stages and of gametocytes, sexual precursor cells crucial for malaria parasite transmission from humans to mosquitoes. We identified three plasmodial SR proteins, PfSRSF12, PfSFRS4 and PfSF-1, which are predominantly present in the nucleus of blood stage trophozoites, PfSRSF12 and PfSF-1 are further detectable in the nucleus of gametocytes. We found that recombinantly expressed SR proteins comprising the Arginine/Serine (RS)-rich domains were phosphorylated by the four PfCLKs in in vitro kinase assays, while a recombinant PfSF-1 peptide lacking the RS-rich domain was not phosphorylated. Since it was hitherto not possible to knock-out the pfclk genes by conventional gene disruption, we aimed at chemical knock-outs for phenotype analysis. We identified five human CLK inhibitors, belonging to the oxo-β-carbolines and aminopyrimidines, as well as the antiseptic chlorhexidine as PfCLK-targeting compounds. The six inhibitors block P. falciparum blood stage replication in the low micromolar to nanomolar range by preventing the trophozoite-to-schizont transformation. In addition, the inhibitors impair gametocyte maturation and gametogenesis in in vitro assays. The combined data show that the four PfCLKs are involved in phosphorylation of SR proteins with essential functions for the blood and sexual stages of the malaria parasite, thus pointing to the kinases as promising targets for antimalarial and transmission blocking drugs.
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, BioXtra, ≥98%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
SAFC
Glycine
Sigma-Aldrich
D-Sorbitol, ≥98%
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Glycine, 99%, FCC
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
D-Sorbitol, 99%
Sigma-Aldrich
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, for molecular biology, ≥98%
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, certified reference material, TraceCERT®