  • Search Results
  • Development of optical probes for in vivo imaging of polarized macrophages during foreign body reactions.
Development of optical probes for in vivo imaging of polarized macrophages during foreign body reactions.
Acta biomaterialia (2014-04-15)
David W Baker, Jun Zhou, Yi-Ting Tsai, Kaitlen M Patty, Hong Weng, Ewin N Tang, Ashwin Nair, Wen-Jing Hu, Liping Tang
ABSTRACT
Plasticity of macrophage (MΦ) phenotypes exist in a spectrum from classically activated (M1) cells, to alternatively activated (M2) cells, contributing to both the normal healing of tissues and the pathogenesis of implant failure. Here, folate- and mannose-based optical probes were fabricated to simultaneously determine the degree of MΦ polarization. In vitro tests show the ability of these probes to specifically target M1 and M2 cells. In an in vivo murine model, they were able to distinguish between the M1-dominated inflammatory response to infection and the M2-dominated regenerative response to particle implants. Finally, the probes were used to assess the inflammatory/regenerative properties of biomaterial implants. Our results show that these probes can be used to monitor and quantify the dynamic processes of MΦ polarization and their role in cellular responses in real time.
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, wood, ≥99%
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
DCC, puriss., ≥99.0% (GC)
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Supelco
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DCC, 99%
Folic acid, European Pharmacopoeia (EP) Reference Standard
USP
Folic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)