  • Search Results
  • Rapid microwave-assisted synthesis of polydextrose and identification of structure and function.
Rapid microwave-assisted synthesis of polydextrose and identification of structure and function.
Carbohydrate polymers (2014-09-27)
Haisong Wang, Yonghui Shi, Guowei Le
ABSTRACT
Microwave irradiation is a rapid and efficient method to synthesize oligomers and can be employed in polysaccharides production. As an artificial polysaccharide, polydextrose is known for its solid performance in food processing and its additional health benefits. This study was aimed at producing polydextrose by microwave irradiation using glucose and sorbitol as substrates; water and phosphoric acid as initiator and catalyst. The actual maximum yield was 99%. Synthetic polydextrose were purified by ethanol elution and Sepherdex G-25 column chromatography. Its purity was demonstrated by the high-performance gel-permeation chromatography as a single symmetrical sharp peak, additionally the average molecular weight was calculated to be 2.131 kDa. FT-IR spectra showed that the synthesized polydextrose has the structural feature similar to Polydextrose-Litesse(®). In vitro fermentation revealed that polydextrose possesses the biological function similar to Polydextrose-Litesse(®) in increasing the concentration of short chain fatty acid and decreasing pH. This research demonstrated the feasibility of a rapid and efficient microwave mediated method to synthesize polydextrose and potentially other value added carbohydrate polymers.
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
D-Sorbitol, 99%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, BioXtra, ≥98%
Sigma-Aldrich
D-Sorbitol, for molecular biology, ≥98%
Sigma-Aldrich
D-Sorbitol, ≥98%
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Supelco
D-(+)-Glucose, analytical standard