  • Search Results
  • An investigation of antitumor efficiency of novel sustained and targeted 5-fluorouracil nanoparticles.
An investigation of antitumor efficiency of novel sustained and targeted 5-fluorouracil nanoparticles.
European journal of medicinal chemistry (2015-02-14)
Van-Minh Le, Jing-Jing Wang, Ming Yuan, The-Long Nguyen, Gui-Fang Yin, Yuan-Hong Zheng, Wei-Bin Shi, Mei-Dong Lang, Lei-Ming Xu, Jian-Wen Liu
ABSTRACT
Traditional chemotherapeutic drugs remain the major treatment for advanced colorectal cancer. However, due to the lack of tumor specificity these drug also destroy healthy tissue and organs, which has been the main reason for treatment failure and mortality. Folate-based drug delivery systems for improving nanoparticle endocytosis have been used to address these problems. Here, folic acid (FA) conjugated mPEG-b-P(CABCL-co-ACL) diblock copolymers were synthesized and characterized by TEM and NMR. Drug loaded nanoparticles were prepared using dialysis method and was obtained with a mean diameter of 45.2 nm with sustained in vitro release profile. In vitro cytotoxicity assay indicated that the cytotoxicity of folate modified nanoparticles were significantly increased compared to free drug and non-folate nanoparticles. In addition, results of hemolytic and histopathologic study suggested that the non-loaded nanoparticle (NL/NP) was non-toxic and biocompatible at the testing concentration. Moreover, in vivo results showed that FA/5-FU/NP effectively inhibited growth of HCT-8 cell-based xenograft tumors in BALB/c mice and revealed stronger antitumor efficacy than other treated groups. Thus, both in vitro and in vivo results exhibited that the folate conjugated mPEG-b-P(CABCL-co-ACL) copolymers have great potential to be used as sustainable and specific colon cancer targeting delivery system for anticancer agents.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Folic acid, European Pharmacopoeia (EP) Reference Standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
USP
Folic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri