  • Search Results
  • Antioxidant and anxiolytic activities of Crataegus nigra Wald. et Kit. berries.
Antioxidant and anxiolytic activities of Crataegus nigra Wald. et Kit. berries.
Acta poloniae pharmaceutica (2014-10-03)
Marija T Popovic-Milenkovic, Marina T Tomovic, Snezana R Brankovic, Biljana T Ljujic, Slobodan M Jankovic
ABSTRACT
Hawthorn has been present for a long time in traditional medicine as antihypertensive, hypolipidemic, anti-inflammatory, gastroprotective, antimicrobial agent. Hawthorn can be used for the cure of stress, nervousness but there is no published paper about actions of Crataegus nigra Wald. et Kit. fruits. The present study was carried out to test free-radical-scavenging and anxiolytic activity of C. nigra fruits. DPPH (alpha,alpha-diphenyl-beta-picrylhydrazyl) assay was used to measure antioxidant activity. BHT, BHA, PG, quercetin and rutin were used as standards. The total amount of phenolic compounds, procyanidins, and flavonoids in the extracts, was determined spectrophotometrically. Results were expressed as equivalents of gallic acid, cyanidin chloride and quercetin equivalents, respectively. LC-MS/MS was used for identification and quantification of phenolic composition. The anxiety effect, expressed as the difference in time spent in the open and closed arms, was measured and compared between groups. Phenolic compound content of Crataegus nigra fruits was 72.7 mg/g. Yield of total flavonoid aglycones was 0.115 mg/g. Procyanidins were 5.6 mg/g. DPPH radical-scavenging capacity of the extracts showed linear concentration dependency, IC50 value were 27.33 microg/mL. Anxiolytic effect was observed. Species Crataegus nigra fruits hydroalcoholic extract showed antioxidant and anxiolytic activity.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Gallic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Guaiacol, oxidation indicator
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material