  • Search Results
  • Interaction of ceruloplasmin with eosinophil peroxidase as compared to its interplay with myeloperoxidase: Reciprocal effect on enzymatic properties.
Interaction of ceruloplasmin with eosinophil peroxidase as compared to its interplay with myeloperoxidase: Reciprocal effect on enzymatic properties.
Free radical research (2015-03-13)
A V Sokolov, V A Kostevich, E T Zakharova, V R Samygina, O M Panasenko, V B Vasilyev
ABSTRACT
Myeloperoxidase (MPO) and eosinophil peroxidase (EPO) are involved in the development of halogenative stress during inflammation. We previously described a complex between MPO and ceruloplasmin (CP). Considering the high structural homology between MPO and EPO, we studied the latter's interaction with CP and checked whether EPO becomes inhibited in a complex with CP. Disc-electrophoresis and gel filtration showed that CP and EPO form a complex with the stoichiometry 1:1. Affinity chromatography of EPO on CP-agarose (150 mM NaCl, 10 mM Na-phosphate buffer, of pH 7.4) resulted in retention of EPO. EPO protects ceruloplasmin from limited proteolysis by plasmin. Only intact CP shifted the Soret band typical of EPO from 413 to 408 nm. The contact with CP likely causes changes in the heme pocket of EPO. Peroxidase activity of EPO with substrates such as guaiacol, orcinol, o-dianisidine, 4-chloro-1-naphtol, 3,3',5,5'-tetramethylbenzidine, and 2,2'-azino-bis(3-ethylbenzthiazoline-6-sulfonate) is inhibited by CP in a dose-dependent manner. Similar to the interaction with MPO, the larger a substrate molecule, the stronger the inhibitory effect of CP upon EPO. The limited proteolysis of CP abrogates its capacity to inhibit the peroxidase activity of EPO. The peptide RPYLKVFNPR (corresponding to amino acids 883-892 in CP) inhibits the peroxidase and chlorinating activity of EPO. Only the chlorinating activity of EPO is efficiently inhibited by CP, while the capacity of EPO to oxidize bromide and thiocyanate practically does not depend on the presence of CP. EPO enhances the p-phenylenediamine-oxidase activity of CP. The structural homology between the sites in the MPO and EPO molecules enabling them to contact CP is discussed.
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine dihydrochloride, tablet, 1 mg substrate per tablet
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Cyanogen bromide, reagent grade, 97%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
SAFC
Glycine
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Cyanogen bromide solution, 5.0 M in acetonitrile
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)