  • Search Results
  • A targeted metabolomics approach toward understanding metabolic variations in rice under pesticide stress.
A targeted metabolomics approach toward understanding metabolic variations in rice under pesticide stress.
Analytical biochemistry (2015-03-15)
Vahideh Mahdavi, Mahdi Moridi Farimani, Fariba Fathi, Alireza Ghassempour
ABSTRACT
Diazinon insecticide is widely applied throughout rice (Oryza sativa L.) fields in Iran. However, concerns are now being raised about its potential adverse impacts on rice fields. In this study, a time-course metabolic change in rice plants was investigated after diazinon treatment using gas chromatography-mass spectrometry (GC-MS), and subsequently the statistical strategy of random forest (RF) was performed in order to find the stress-associated effects. According to the results, a wide range of metabolites were dynamically varied as a result of the plant response to diazinon such as biosynthesis and metabolism of sugars, amino acids, organic acids, and phenylpropanoids, all correlating with the exposure time. Plant response was involved in multiple metabolic pathways, most of which were correlated with the exposure time. In this study, RF was explored as a potential multivariate method for GC-MS analysis of metabolomics data of rice (O. sativa L.) plants under diazinon stress; more than 31 metabolites were quantitatively determined, and time-course metabolic response of the plant during different days after treatment was measured. Results demonstrated RF as a potential multivariate method for GC-MS analysis of changes in plant metabolome under insecticide stress.
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
Trifluoroacetamide, 97%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
D-Sorbitol, BioXtra, ≥98%
Sigma-Aldrich
D-Sorbitol, ≥98%
Sigma-Aldrich
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, for molecular biology, ≥98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methoxylamine hydrochloride solution, 25-30 wt. % in H2O
Sigma-Aldrich
D-Sorbitol, 99%
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Methoxyamine hydrochloride, 98%
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Methanol, NMR reference standard