  • Search Results
  • Ultraviolet-C light inactivation of Escherichia coli O157:H7 and Listeria monocytogenes on organic fruit surfaces.
Ultraviolet-C light inactivation of Escherichia coli O157:H7 and Listeria monocytogenes on organic fruit surfaces.
International journal of food microbiology (2015-07-01)
Achyut Adhikari, Roopesh M Syamaladevi, Karen Killinger, Shyam S Sablani
ABSTRACT
This study investigated UV-C light inactivation of Escherichia coli O157:H7 and Listeria monocytogenes on the surface of organic apples, pears, strawberries, red raspberries and cantaloupes. Fruit surfaces spot inoculated with cocktail strains of E. coli O157:H7 and L. monocytogenes were exposed to UV-C doses up to 11.9 kJ/m(2) at 23 °C. Fruit surface roughness, contact angle, and surface energy were determined and correlated with UV-C inactivation kinetics. Results demonstrate that bacterial pathogens on fruit surfaces respond differently to UV-C light exposure. E. coli O157:H7 on apple and pear surfaces was reduced by 2.9 and 2.1 log CFU/g, respectively when treated with UV-C light at 0.92 kJ/m(2) (60s). For berries, the reduction of E. coli O157:H7 was lower with 2.0 (strawberry) and 1.1 log CFU/g (raspberry) achieved after UV-C treatment at 7.2 kJ/m(2) (8 min) and at 10.5 kJ/m(2) (12 min), respectively. Similarly, a higher reduction of L. monocytogenes was observed on apple (1.6 log CFU/g at 3.75 kJ/m(2)) and pear (1.7 log CFU/g at 11.9 kJ/m(2)) surfaces compared to cantaloupe and strawberry surfaces (both achieved 1.0 log CFU/g at 11.9 kJ/m(2)). L. monocytogenes shows more resistance than E. coli O157:H7. Inactivation rates were higher for less hydrophobic fruits with smoother surfaces (apples and pears) as compared to fruits with rougher surfaces (cantaloupe, strawberry and raspberry). Findings indicate that UV-C light can effectively reduce E. coli O157:H7 and L. monocytogenes populations on fruit and berry surfaces. However, surface characteristics influence the efficacy of UV-C light.
Sigma-Aldrich
6-Benzylaminopurine hydrochloride, suitable for plant cell culture, ≥98%
Sigma-Aldrich
D-Sorbitol, BioXtra, ≥98%
Sigma-Aldrich
D-Sorbitol, ≥98%
Sigma-Aldrich
D-Sorbitol, for molecular biology, ≥98%
Sigma-Aldrich
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, 99%
Sigma-Aldrich
Diiodomethane, ReagentPlus®, 99%, contains copper as stabilizer
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)