  • Search Results
  • Characterization of a cypermethrin-degrading Methylobacterium sp. strain A-1 and molecular cloning of its carboxylesterase gene.
Characterization of a cypermethrin-degrading Methylobacterium sp. strain A-1 and molecular cloning of its carboxylesterase gene.
Journal of basic microbiology (2015-07-02)
Corinna Diegelmann, Joachim Weber, Regina Heinzel-Wieland, Michael Kemme
ABSTRACT
A novel mesophilic bacterial strain, designated A-1, was isolated from microbially contaminated biopolymer microcapsules. The bacterium was able to withstand and grow in liquid cultures supplemented with the pyrethroid cypermethrin in concentrations up to 400 mg L(-1) . Furthermore, strain A-1 could use cypermethrin as sole carbon source and could degrade >50% of it in 12 h. Based on phenotypic and chemotaxonomic characterization, and phylogenetic analysis of 16S rRNA gene sequence, the strain A-1 was identified as Methylobacterium sp., which is the first reported cypermethrin degrader of methylotrophic bacteria. A role for esterase activity in cypermethrin biodegradation was presumed. Therefore, the carboxylesterase gene mse1 was amplified from the Methylobacterium sp. strain A-1 genome and the resulting 1 kb amplicon cloned into E. coli. Sequence analysis of the mse1-DNA insert revealed an open reading frame of 633 bp encoding for a putative carboxylesterase of 210 amino acid residues with a predicted molecular mass of 22 kDa. The amino acid sequence of the deduced enzyme MsE1 with the catalytic triad Ser106 , Asp156 , and His187 was found to be similar to that of α/β-hydrolase fold proteins. The active site Ser106 residue is located in the consensus pentapeptide motif Gly-X-Ser-X-Gly that is typical of esterases.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
1,2,4,5-Tetramethylbenzene, 98%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free