  • Search Results
  • Impact of Infection Dose and Previous Serum Antibodies against the Locus of Enterocyte Effacement Proteins on Escherichia coli O157:H7 Shedding in Calves following Experimental Infection.
Impact of Infection Dose and Previous Serum Antibodies against the Locus of Enterocyte Effacement Proteins on Escherichia coli O157:H7 Shedding in Calves following Experimental Infection.
BioMed research international (2015-07-15)
L Martorelli, C J Hovde, D A Vilte, A Albanese, E Zotta, C Ibarra, R J C Cantet, E C Mercado, A Cataldi
ABSTRACT
Escherichia coli O157:H7 is the main causative agent of haemolytic uremic syndrome. Cattle are the main reservoir of these bacteria, and have been shown to develop immune response to colonization. Our aim was to investigate the faecal shedding pattern of E. coli O157:H7 in calves challenged intragastrically with either 10(8) or 10(10) CFU, as well as the ability of specific preexisting antibodies to reduce shedding of the pathogen. Shedding was analysed by direct counting as well as enrichment of rectoanal mucosal swabs. Statistical analysis was performed using a linear model for repeated measures with and without the inclusion of preexisting antibodies against the carboxy-terminal fraction of intimin-γ (γ-intimin C280) as a covariable. Results suggest that there is a statistical difference in the area under the shedding curves between both doses for 14 as well as 28 days after challenge (p = 0.0069 and 0.0209, resp.). This difference is increased when the prechallenge antibodies are taken into account (p = 0.0056 and 0.0185). We concluded that the bacterial dose influences shedding on calves experimentally challenged and that preexisting antibodies against E. coli O157:H7 γ-intimin C280 could partially reduce faecal excretion.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Nalidixic acid, ≥98%
Sigma-Aldrich
D-Sorbitol, BioXtra, ≥98%
Sigma-Aldrich
D-Sorbitol, ≥98%
Sigma-Aldrich
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, for molecular biology, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
D-Sorbitol, 99%
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri