  • Search Results
  • Suitable coating material for microencapsulation of spray-dried fish oil.
Suitable coating material for microencapsulation of spray-dried fish oil.
Journal of food science and technology (2015-07-04)
M Tirgar, S Jinap, I S M Zaidul, H Mirhosseini
ABSTRACT
This study was conducted to screen the most suitable coating material for the production of microencapsulated fish oil powder using ternary blends of maltodextrin (15, 25 % w/w), Arabic gum (2.5, 7.5 % w/w), and methylcellulose (0.5, 1.5 % w/w). The physical properties of fish oil emulsion and encapsulated powders were evaluated. Arabic gum (5 % w/w) showed the most significant (p < 0.05) effect on the surface mean diameter of the droplets in the emulsion. Maltodextrin had the most significant (p < 0.05) effect on the centrifuge stability of the emulsion and the amount of surface oil of the powder at 15 and 20 % (w/w) respectively, whereas methylcellulose (0.5 % w/w) had the most significant (p < 0.05) effect on the width distribution of the droplets in the emulsion. The total optimal area leading to the formation of coating material with desirable physical properties was expected to be obtained by the combination of 16 % (w/w) maltodextrin, 6.5 % (w/w) Arabic gum, and 0.88 % (w/w) methylcellulose respectively.
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)