Merck
Role of PTEN in neutrophil extracellular trap formation.
Molecular immunology (2015-04-29)
Shahram Teimourian, Ehsan Moghanloo
ABSTRACT
NETosis has been associated with a particular mode of cell death although it is still controversial as to what extent autophagy is involved in NETosis. Class I/AKT/mTOR pathway is a key regulator of autophagy. PTEN tumor suppressor gene encodes a dual specificity phosphatase that antagonizes the phosphatidylinositol 3-kinase in class the I/AKT/mTOR pathway. In this study, we investigated the effects of PTEN down-regulation as well as overexpression on NETosis. Our results show that 35% of HL-60 differentiated neutrophil-like cells generated NETs by PMA. The portion of the population that produced NETs in PTEN knockdown HL-60 differentiated neutrophils was 9% and in PTEN overexpressed HL-60 differentiated neutrophils, it was 56%. Our results show that increasing PTEN expression increases NETs formation in neutrophils, and its suppression reduces NETs.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human PTEN