  • Search Results
  • Insights into the influence of the cooling profile on the reconstitution times of amorphous lyophilized protein formulations.
Insights into the influence of the cooling profile on the reconstitution times of amorphous lyophilized protein formulations.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-08-09)
Karen E Beech, James G Biddlecombe, Christopher F van der Walle, Lee A Stevens, Sean P Rigby, Jonathan C Burley, Stephanie Allen
ABSTRACT
Lyophilized protein formulations must be reconstituted back into solution prior to patient administration and in this regard long reconstitution times are not ideal. The factors that govern reconstitution time remain poorly understood. The aim of this research was to understand the influence of the lyophilization cooling profile (including annealing) on the resulting cake structure and reconstitution time. Three protein formulations (BSA 50mg/ml, BSA 200mg/ml and IgG1 40mg/ml, all in 7% w/v sucrose) were investigated after cooling at either 0.5°C/min, or quench cooling with liquid nitrogen with/without annealing. Significantly longer reconstitution times were observed for the lower protein concentration formulations following quench cool. Porosity measurements found concomitant increases in the surface area of the porous cake structure but a reduction in total pore volume. We propose that slow reconstitution results from either closed pores or small pores impeding the penetration of water into the lyophilized cake.
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%