Merck
LC/MS/MS Analysis of Pesticide Residues in Pistachios on the Ascentis® Express RP-Amide Column after QuEChERS Extraction
LC/MS/MS Analysis of Pesticide Residues in Pistachios on the Ascentis® Express RP-Amide Column after QuEChERS Extraction application for LC-MS, application for SPE
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2 μm UHPLC Column
2 μm particle size, L × I.D. 10 cm × 2.1 mm
SPE tube or plate
Product No.
Description
Pricing
Supel QuE
Z-Sep+ Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
accessory
Product No.
Description
Pricing
Certified Vial Kit, Low Adsorption (LA), 2 mL, pk of 100
volume 2 mL, amber glass vial (with marking spot), natural PTFE/silicone septa (with slit), thread for 9 mm
sample prep
Product No.
Description
Pricing
Supel QuE
Citrate Extraction Tube, pk of 50, suitable for EN 15662
Supel QuE
Empty Centrifuge Tube with Lid, centrifuge tube volume 50 mL, pk of 50, suitable for EN 15662:2008 per BS
Benchmixer XL multi-tube vortexer
AC/DC input 115 V AC
Supel QuE
Z-Sep+ Tube, centrifuge tube volume 2 mL, pack of 100 ea
QuEChERS Shaker and Rack Starter Kit
AC/DC input 115 V AC (USA Compatible Plug)
CONDITIONS
sample/matrix
5 g ground pistachios spiked with pesticides at 10 ng/g
extraction process
add 10 mL water and 10 mL acetonitrile; shake for 1 hour at 2500 rpm; add contents of Supel QuE citrate extraction tube; shake for 1 minute; centrifuge at 5000 rpm fo 5 minutes; draw off supernatant.
clean-up tube
Supel QuE Z-Sep + (55408-U)
clean-up process
transfer 1 mL of extract to 2 mL tube containing 75 mg Z-Sep+; shake for 1 minute; centrifuge at 5000 rpm for 3 minutes; draw off supernatant.
sample preparation
Dilute 100 μL of cleaned extract to a final volume of 1 mL in 5 mM ammonium formate/0.1% formic acid in water. Transfer diluted sample to vial for LC/MS/MS analysis
column
Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2 μm particles (51576-U)
mobile phase
[A] 5 mM ammonium formate + 0.1% formic acid in water; [B] 5 mM ammonium formate + 0.1% formic acid in 95:5 acetonitrile:H2O
gradient
95% A from 0 to 1 min; to 100% B in 13 min; held at 100% B for 1.5 min; to 95% A in 0.5 min; held at 95% A for 1.5 min
flow rate
0.4 mL/min
column temp.
30 °C
detector
MS, ESI (+), MRM
injection
5 μL
extraction tube
PK50 Supel QuE Citrate (EN) Tube (55227-U)
Description
Analysis Note
The quick, easy, cheap, effective, rugged, and safe (QuEChERS) method can be used to analyze multiple pesticide residues found in pistachios. Because they contain a significant amount of fat, a cleanup step should be used as part of the QuEChERS method. In this study, QuEChERS extraction and cleanup using Z-Sep+ was used for the analysis of pesticide residues of interest in pistachios. Average recoveries obtained for replicate samples spiked at 10 ng/g were in the range of 70-120% for 23 of the 25 pesticides analyzed. .
Other Notes
Freeze whole pistachios (with shells) using liquid nitrogen, and grind.
Legal Information
Ascentis is a registered trademark of Sigma-Aldrich Co. LLC