radiello® Diffusive Air Sampler Calibration Solutions and Kits
radiello Table of Contents
Calibration Solution for H2S
Product No. RAD171
RAD171 relieves you from the task of preparing the sodium sulfide standard solution for the calibration curve used for the determination of H2S by the cartridge RAD170.
Since sodium sulfide is deliquescent, its weight is not a primary standard and sodium sulfide solution need titration once prepared. Moreover, titration must be repeated often due to the instability of diluted solution (one hour time is sufficient to decrease sulfide content by 10%).
RAD171 is a methylene blue concentrated solution that, once diluted 1:50, provides the same absorbance value at 665 nm of a sodium sulfide solution of with concentration 1.145 μg·ml-1 sulfide ions. This concentration value has been chosen to obtain the highest absorbance value within the linearity range of the spectrophotometer.
To obtain a complete calibration curve, just dilute the mother solution as shown in the table.
RAD171 contains 100 mL solution that allows you to prepare as many as 50 calibration curves. Kept closed at room temperature, code 171 solution is stable for at least one year.
Calibration Solution for Aldehydes
Product No. RAD302
Calibration curves for aldehydes are obtained with standard solutions of the corresponding 2,4-dinitrophenylhydrazones. Although their synthesis is straightforward, their purification is tricky and time-consuming.
RAD302 offers a certified and convenient choice: a solution of nine 2,4-dinitrophenylhydrazones in a solvent compatible with HPLC eluents and with concentrations suitable for the preparation of calibration curves in the range usually spanned by radiello samples.
RAD302 is delivered as 10 mL of acetonitrile solution of the nine 2,4-dinitrophenylhydrazones formed by the aldehydes listed in the table, contained in a pierceable-septum crimped cap vial. The listed concentration values are indicative, actual ones are certified for each lot.
Kept tightly capped in a dark place at 4 °C, the solution is stable for at least four months.
Calibration Kit for BTEX (CS2 desorption)
Product No. RAD405
This calibration kit has been conceived for the analysis of BTEX sampled in urban environments by the cartridge RAD130 and chemically desorbed by carbon disulfide.
The kit may be used both for routine calibration and for scheduled quality control of the calibration procedure described above.
The kit is composed of 12 RAD130 cartridges, three of which are blanks and nine, divided into three groups of three, preloaded with BTEX to simulate 7 days exposures (10,080 minutes) to the concentrations listed in the table. The values shown are indicative, and actual ones are certified for each lot.
The mass of each analyte deposited onto the cartridge spans the whole range of concentrations usually found in urban environments, extreme values included. BTEX loading is performed by injection of precisely known amounts of vaporized standard solutions in CS2 of the five compounds under nitrogen flow.
Kept at 4 °C, the cartridges are stable for at least four months.
Calibration Kit for VOCs in Workplace Environments
Product No. RAD406
This kit has been conceived for scheduled quality control of the calibration procedure for the analysis of volatile organic compounds (VOCs) sampled by RAD130 cartridges in workplace environments.
It is composed of 12 RAD130 cartridges, three of which are blanks and nine, divided into three groups of three, preloaded with VOCs to simulate 8 hours exposures (480 minutes) to the concentrations listed in the table. The values shown are indicative, actual ones are certified for each lot.
The composition of the mixture is simple but it includes compounds with different polarity. The loaded mass is calculated in order to represent exposures to 0.5, 1 and 2 times the TLV value for the mixture. VOCs loading is performed by injection of precisely known amounts of calibrated mixtures of the eight compounds under nitrogen flow.
Kept at 4 °C, the cartridges are stable for at least four months.
Calibration Kit for BTEX (thermal desorption)
Product No. RAD407
This calibration kit has been conceived for the analysis of BTEX sampled in urban environments by the cartridge RAD145 and thermally desorbed.
The kit may be used both for routine calibration and for scheduled quality control of the calibration procedure described on page E5.
It is composed of 12 RAD145 cartridges, three of which are blanks and nin, divided into three groups of three, preloaded with BTEX to simulate 7 days exposures (10,080 minutes) to the concentrations listed in the table. The values shown are indicative, actual ones are certified for each lot.
BTEX loading is performed by injection of precisely known amounts of vaporized standard solutions in methanol of the five compounds under nitrogen flow. During the analysis the chromatographic peak of methanol will be visible.
Kept at 4 °C, the cartridges are stable for at least four months.