Merck
HomePhotovoltaics & Solar CellsNon-Fullerene Acceptors for Organic Photovoltaic Applications
Non-Fullerene Acceptors for Organic Photovoltaic Applications
François Grenier , Amélie Robitaille
Brilliant Matters Organic Electronics Inc.
Organic photovoltaics
The emerging organic photovoltaic (OPV) technology is very promising for low-cost solar energy production. OPV devices can be produced using high-throughput, large-volume printing methods on lightweight and flexible plastic substrates, making them easy to deploy and use in innovative ways. For example, they can be integrated into buildings and windows, used as architectural design, in wearable applications, etc.
However, a major challenge has been how to increase the power conversion efficiency (PCE) of OPV devices to reach values comparable to well-established, mass-produced solar cells (ex: multicrystalline silicon can reach 22.3% in the lab).1 Until recently, the most efficient semiconductors used in OPV were a combination of p-type polymer (ex: PTB7-Th 794333, PBDB-T 901099, PffBT4T-2OD 900720) with n-type fullerene derivative (such as PC61BM 684457, PC71BM 684465, ICBA 745839).2 This allowed the PCE to reach values close to 11%, a value believed to be close to the theoretical limit for polymer/fullerene single layer OPV devices.3
A paradigm shift
Fortunately, a recent discovery has overcome the limited PCE performance, making OPV a serious contender in emerging solar energy technologies. A new class of n-type molecular materials has improved the PCE to 16% for single-junction devices.4,19 These new materials, broadly labeled as “non-fullerene acceptors’’ (NFA), offer several advantages over fullerene acceptors:
  • increased photocurrent due to a higher absorption coefficient
  • improved cell voltage due to a reduction in non-radiative energy losses5
  • high device photostability6,20 .
Theoretical estimates are now more optimistic for NFA-based OPV devices and it is believed efficiencies over 19% are realistically achievable by optimising polymer/NFA bulk heterojunctions.5
A new molecular design
Most of the high performance NFAs share common molecular features that allow them to outperform fullerene and other materials. As often seen in conjugated polymers, they feature a combination of electron-rich and electron-poor conjugated moieties to lower the bandgap and achieve a broader absorption of the solar spectrum. However, what sets them apart is that only the electron-poor side-groups are accessible to interact with the p-type material because the electron-rich core is shielded by bulky side-groups (Figure 1). This approach is thought to improve the driving force to create separate charges from an exciton.5 As a result, it is possible to achieve higher voltages in photovoltaic devices than with fullerene derivatives. Combined with an increased absorption coefficient and photocurrent generation, NFAs are the new n-type materials of choice for high-performance organic solar cells.
Typical features of the current high performance NFAs
Figure 1. Typical features of the current high performance NFAs (figure adapted from the literature).5 1. Compact intermolecular 𝝅-𝝅 overlap (intermolecular charge transport channel); 2. Conjugated backbone shielded by alkyls (intramolecular charge transport channel); 3. Non-shielded electron pulling group (to form charge transfer chaneel with donors).
Versatile framework for optimized performance
Another benefit of NFAs in organic photovoltaics is they can easily be chemically modified to change their electronic properties, their optical bandgap, their crystallinity and their solubility. Several classes of materials have proven their ability to obtain high-performance photovoltaic devices with a variety of different p-type materials. Some example are the ITIC7 (900799, 901423, 900947, 906387, 900803, 900800), BTP4,19 (Y6 or BTP-4F 908509), IEICO8 and IDTBR9 (900810, 900853, 910635) derivatives shown in Figure 2.10
Examples of high-performance NFAs.
Figure 2.Examples of high-performance NFAs.
It is possible to fine-tune the electronic properties of NFAs by adding substituents on the end-groups. For example, halogens and methyl moieties can increase or decrease their energy levels and bandgap, which is a useful method to maximize cell performance. These substituents also influence the crystallinity of NFAs, an important factor to control phase separation, blend morphology and device lifetime.6
UV-vis spectra and energy levels
UV-vis spectra and energy levels of ITIC and energy levels of BTP and IEICO derivatives.
UV-vis spectra and energy levels of ITIC and energy levels of BTP and IEICO derivatives.
Figure 3. UV-vis spectra and energy levels of ITIC and energy levels of BTP and IEICO derivatives.
Using NFAs in organic solar cells
NFAs can be used to make organic photovoltaic devices using the same methods developed for fullerene n-type molecules. They can be solubilized in the same solvents commonly used to deposit the bulk-heterojunction layer, for example chloroform, chlorobenzene, dichlorobenzene, xylenes, etc.
As with fullerene derivatives, optimization of the morphology and phase separation can be investigated using small amounts of high boiling point solvent additives, for example 1,8-diiodooctane, 1-chloronaphtalene and anisaldehyde. Lower boiling point co-solvents such as for example o-xylene and N-methylpyrrolidone have also been successfully used with NFAs. Moreover, solvent vapor and thermal annealing can be used to optimize device performance. Please refer to the excellent review by Gurney et al. for more information about morphology control of polymer/NFAs blends.11
Applications and markets
The printability and versatility of NFAs allow them to be used in innovative photovoltaic devices as well as other technologies. For example, wide-bandgap NFAs (> 1.75 eV, ex: IDT-2BR 910635, PBDB-T-SF 906344, and PBDB-T-2Cl 906352) are excellent candidates for low-light applications (indoor lighting) to power IoT devices, as they yield higher cell voltages and absorb most of the photons emitted by artificial light.12 Using low-bandgap materials, on the other hand, (ex: Y6 or BTP-4F 908509, BTP-4Cl, IEICO-4Cl, and BT-CIC 906824) is desirable for semi-transparent applications (ex: windows integration) as photon absorption primarily occurs in the near-infrared spectrum, with weaker extinction coefficients in the visible spectrum.13 Building integration is also an important market for OPV. Having access to NFAs with various absorption spectra, and consequently different colors, can be beneficial to make aesthetically pleasing solar cells integrated into architectural designs.
Applications for NFAs are not limited to organic solar cells. Their reduced bandgap compared to fullerene derivatives make them well-suited for making flexible, printed, near-infrared photodetectors with improved sensitivity.14 They can also be employed to make organic thin-film transistors15 and have potential in printed organic memory devices.16
A brilliant future for organic photovoltaics
The innovative molecular design of NFAs allows organic photovoltaics to overcome the performance limitation of OPV technology and to reach higher efficiencies. Power conversion efficiencies >16% in single layer devices can now be achieved4 and a PCE over 19% is expected to be reachable using polymer/NFA bulk heterojunctions.5 These materials offer improved photon absorption, reduced energy losses and versatility to offer new possibilities for device engineering. Adding new interlayers, optical adjustment layers17 and developing new device configurations18 could lead to even higher values, making NFAs one of the most important recent innovations for the OPV research field and market, and for renewable energy production at large.
Materials
Loading
References
1.
Best Research-Cell Efficiency Chart (accessed August 23). [Internet]. Available from: https://www.nrel.gov/pv/cell-efficiency
2.
Jung JW, Jo JW, Jung EH, Jo WH. 2016. Recent progress in high efficiency polymer solar cells by rational design and energy level tuning of low bandgap copolymers with various electron-withdrawing units. Organic Electronics. 31149-170. http://dx.doi.org/10.1016/j.orgel.2016.01.034
3.
Scharber M, Mühlbacher D, Koppe M, Denk P, Waldauf C, Heeger A, Brabec C. 2006. Design Rules for Donors in Bulk-Heterojunction Solar Cells?Towards 10?% Energy-Conversion Efficiency. Adv. Mater.. 18(6):789-794. http://dx.doi.org/10.1002/adma.200501717
4.
Fan B, Zhang D, Li M, Zhong W, Zeng Z, Ying L, Huang F, Cao Y. 2019. Achieving over 16% efficiency for single-junction organic solar cells. Sci. China Chem.. 62(6):746-752. http://dx.doi.org/10.1007/s11426-019-9457-5
5.
Hou J, Inganäs O, Friend RH, Gao F. 2018. Organic solar cells based on non-fullerene acceptors. Nat. Mater.. 17(2):119-128. http://dx.doi.org/10.1038/nmat5063
6.
Du X, Heumueller T, Gruber W, Classen A, Unruh T, Li N, Brabec CJ. 2019. Efficient Polymer Solar Cells Based on Non-fullerene Acceptors with Potential Device Lifetime Approaching 10 Years. Joule. 3(1):215-226. http://dx.doi.org/10.1016/j.joule.2018.09.001
7.
Cui Y, Yao H, Hong L, Zhang T, Xu Y, Xian K, Gao B, Qin J, Zhang J, Wei Z, et al. 2019. Achieving Over 15% Efficiency in Organic Photovoltaic Cells via Copolymer Design. Adv. Mater.. 31(14):1808356. http://dx.doi.org/10.1002/adma.201808356
8.
Xiao M, Zhang K, Dong S, Yin Q, Liu Z, Liu L, Huang F, Cao Y. 2018. High-Performance Ternary Nonfullerene Polymer Solar Cells with Both Improved Photon Harvesting and Device Stability. ACS Appl. Mater. Interfaces. 10(30):25594-25603. http://dx.doi.org/10.1021/acsami.8b06822
9.
Baran D, Gasparini N, Wadsworth A, Tan CH, Wehbe N, Song X, Hamid Z, Zhang W, Neophytou M, Kirchartz T, et al. 2018. Robust nonfullerene solar cells approaching unity external quantum efficiency enabled by suppression of geminate recombination. Nat Commun. 9(1): http://dx.doi.org/10.1038/s41467-018-04502-3
10.
Zhang G, Zhao J, Chow PCY, Jiang K, Zhang J, Zhu Z, Zhang J, Huang F, Yan H. 2018. Nonfullerene Acceptor Molecules for Bulk Heterojunction Organic Solar Cells. Chem. Rev.. 118(7):3447-3507. http://dx.doi.org/10.1021/acs.chemrev.7b00535
11.
Gurney RS, Lidzey DG, Wang T. 2019. A review of non-fullerene polymer solar cells: from device physics to morphology control. Rep. Prog. Phys.. 82(3):036601. http://dx.doi.org/10.1088/1361-6633/ab0530
12.
Cui Y, Wang Y, Bergqvist J, Yao H, Xu Y, Gao B, Yang C, Zhang S, Inganäs O, Gao F, et al. 2019. Wide-gap non-fullerene acceptor enabling high-performance organic photovoltaic cells for indoor applications. Nat Energy. 4(9):768-775. http://dx.doi.org/10.1038/s41560-019-0448-5
13.
Cui Y, Yang C, Yao H, Zhu J, Wang Y, Jia G, Gao F, Hou J. 2017. Efficient Semitransparent Organic Solar Cells with Tunable Color enabled by an Ultralow-Bandgap Nonfullerene Acceptor. Adv. Mater.. 29(43):1703080. http://dx.doi.org/10.1002/adma.201703080
14.
Lee J, Ko S, Lee H, Huang J, Zhu Z, Seifrid M, Vollbrecht J, Brus VV, Karki A, Wang H, et al. 2019. Side-Chain Engineering of Nonfullerene Acceptors for Near-Infrared Organic Photodetectors and Photovoltaics. ACS Energy Lett.. 4(6):1401-1409. http://dx.doi.org/10.1021/acsenergylett.9b00721
15.
Yang F, Li C, Lai W, Zhang A, Huang H, Li W. Halogenated conjugated molecules for ambipolar field-effect transistors and non-fullerene organic solar cells. Mater. Chem. Front.. 1(7):1389-1395. http://dx.doi.org/10.1039/c7qm00025a
16.
Zhou L, Mao J, Ren Y, Han S, Roy VAL, Zhou Y. 2018. Recent Advances of Flexible Data Storage Devices Based on Organic Nanoscaled Materials. Small. 14(10):1703126. http://dx.doi.org/10.1002/smll.201703126
17.
Shen P, Long Y, Wang G, Wang Y, Guo W, Shen L. Semi-transparent polymer solar cells with optical adjusting layers. J. Mater. Chem. C. 6(35):9494-9500. http://dx.doi.org/10.1039/c8tc03289k
18.
Kaake LG. 2019. Towards the Organic Double Heterojunction Solar Cell. Chem. Rec.. 19(6):1131-1141. http://dx.doi.org/10.1002/tcr.201800180
19.
Cui Y, Yao H, Zhang J, Zhang T, Wang Y, Hong L, Xian K, Xu B, Zhang S, Peng J, et al. 2019. Over 16% efficiency organic photovoltaic cells enabled by a chlorinated acceptor with increased open-circuit voltages. Nat Commun. 10(1): http://dx.doi.org/10.1038/s41467-019-10351-5
20.
Holliday S, Ashraf RS, Wadsworth A, Baran D, Yousaf SA, Nielsen CB, Tan C, Dimitrov SD, Shang Z, Gasparini N, et al. 2016. High-efficiency and air-stable P3HT-based polymer solar cells with a new non-fullerene acceptor. Nat Commun. 7(1): http://dx.doi.org/10.1038/ncomms11585