CDS003086

Sigma-Aldrich

6-Bromohexanamide

AldrichCPR

Empirical Formula (Hill Notation):
C6H12BrNO
Numéro CAS:
Poids moléculaire:
194.07
Numéro MDL:
ID de substance PubChem:

SMILES string

NC(=O)CCCCCBr

InChI

1S/C6H12BrNO/c7-5-3-1-2-4-6(8)9/h1-5H2,(H2,8,9)

InChI key

LGCGXHIRLZORQA-UHFFFAOYSA-N

Autres remarques

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

storage_class_code

11 - Combustible Solids

WGK Allemagne

WGK 3

Flash Point(F)

Not applicable

Point d'éclair C

Not applicable

Certificat d'analyse

Certificat d'origine

Amika Mori et al.
Pharmaceutics, 13(1) (2021-01-13)
The structure-activity relationship of mono-ion complexes (MICs) for plasmid DNA (pDNA) delivery by muscular injection is demonstrated. MICs were formed between pDNA and monocationic poly(ethylene glycol) (PEG) macromolecules. As monocationic PEGs, the ω-amide-pentylimidazolium (APe-Im) end-modified PEGs with a stable amide...

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique