SYX00398

Sigma-Aldrich

4-(Aminomethyl)-2-isopropylthiazole dihydrochloride

Empirical Formula (Hill Notation):
C7H12N2S · 2HCl
Numéro CAS:
Poids moléculaire:
229.17
Numéro MDL:
ID de substance PubChem:

SMILES string

NCC1=CSC(C(C)C)=N1.Cl.Cl

InChI

1S/C7H12N2S.2ClH/c1-5(2)7-9-6(3-8)4-10-7;;/h4-5H,3,8H2,1-2H3;2*1H

InChI key

FFYQRDDCOHMJTM-UHFFFAOYSA-N

Autres remarques

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Skull and crossbones

Mention d'avertissement

Danger

Mentions de danger

Conseils de prudence

hazcat

Acute Tox. 3 Oral

storage_class_code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK Allemagne

WGK 3

Flash Point(F)

Not applicable

Point d'éclair C

Not applicable

Certificat d'analyse

Certificat d'origine

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique