Z247715
Coors table-type removable plate Buchner funnel
Perforated plate
NACRES:
NB.43
Matériau
porcelain
manufacturer/tradename
Coorstek 60264
L × l × H
19 cm (7 1/2  in.) × 7.6 cm (3 in.) × 12 cm (4 3/4  in.)
Poids
0.8 lb
Vous recherchez des produits similaires ? Visit Guide de comparaison des produits
Description générale
The Scienceware® Buchner funnel trowel is specifically designed for use with 18, 24 and 36 in. Buchner funnels. Just apply slight downward pressure and ′sweep′ the trowel along the surface of the precipitate to quickly and efficiently smooth fissures appearing during the filtration process.
  • Large surface area smooths fissures faster than traditional narrow spatula method
  • Dual-beveled edges for bi-directional smoothing
  • Curved ends match Buchner funnel sidewalls allowing easy access to entire cake surface
  • FDA-grade high-density polyethylene (HDPE) complies with industry standards and provides maximum chemical-resistance
  • Welded construction with no hollow cavities assures hygienic use and easy cleaning
  • Compatible with industrial dishwasher temperatures and cleaners
Porcelain. Removable perforated plate allows thorough cleaning. Side-outlet for vacuum. Glazed inside and out except for rim and seat portion of plate and funnel. Order funnel and perforated plate separately.
Informations légales
Coors is a trademark of Coorstek, Inc.
Scienceware is a registered trademark of SP Scienceware, a division of SP
Certificat d'analyse
Certificat d'origine
Contacter notre Service technique
Réseaux sociaux
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Recherche. Développement. Production
Nous sommes un des tout premiers fournisseurs du secteur mondial des Sciences de la vie proposant des solutions et des services pour la recherche, le développement et la production en biotechnologies, et le développement et la production de produits thérapeutiques pharmaceutiques.
© 2021 Merck KGaA, Darmstadt, Allemagne et/ou ses sociétés affiliées. Tous droits réservés.
La reproduction d'une quelconque partie du contenu de ce site est strictement interdite sans autorisation.