ZMQSP0D02
Milli-Q
Q-POD® Stand for Remote Dispensing
Convenient remote ultrapure water stand
Conditionnement
pkg of 1 unit
manufacturer/tradename
Q-POD®
H × D × diam.
22.8 in. × 9 in. × 9 in.
Compatibilité
for use with Milli-Q® Direct
for use with Milli-Q® Reference
Description générale
The Q-POD® Remote Stand can be installed up to 2 meters from the purification system. This allows for convenient ultrapure water dispensing adapted to the laboratory configuration and users′ specific needs (on the bench, close to the sink, …).
For use with Milli-Q® Reference and Milli-Q® Direct water purification systems.
Caractéristiques et avantages
The ability to adapt to the laboratory configuration allows flexibility to create a more efficient working environment.
Informations légales
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-POD is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificat d'analyse
Certificat d'origine
Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..
Contacter notre Service technique