HLMIR1421

Sigma-Aldrich

MISSION® Lenti microRNA, Human

hsa-miR-4435

NACRES:
NA.51

Niveau de qualité

200

Gamme de produits

MISSION®

Concentration

≥1x106 VP/ml (via p24 assay)

Séquence mature

AUGGCCAGAGCUCACACAGAGG

N° d'accession Sanger mature/mineur

N° d'accession Sanger microARN

Expédié(e)(s) dans

dry ice

Temp. de stockage

−70°C

Description générale

Sigma′s Mission Lenti-miRs express miRNAs from a common backbone, whose structure meets requirements for accurate Dicer processing and a partially complementary strand is designed to mimic the base pairing pattern in the backbone structure using a proprietary algorithm. Oligos containing the microRNA sequences are cloned into the TRC2-pLKO-puro vector. Each miRNA construct has been cloned and sequence verified. Mature microRNA sequences are obtained from miRBase.

Lentiviral transduction particles are produced from sequence-verified lentiviral plasmid vectors. Oligos containing the microRNA sequences are cloned into the TRC2-pLKO-puro vector. Co-transfection of this vector into the appropriate cell line with compatible packaging plasmids produces viral particles that can be used to transduce mammalian cells. The polymerase II promoter, elongation factor 1 alpha (EF1A), was chosen to drive miRNA expression needed for reverse transcription of viral RNA and integration of viral DNA into the host cell genome. Additionally, the Woodchuck Hepatitis Post-Transcriptional Regulatory element allowing for enhanced expression of transgenes delivered by lentiviral vectors. This lentiviral vector also carries a puromycin resistance gene for selection of cells. Unlike murine-based MMLV or MSCV retroviral systems, lentiviral-based particles permit efficient infection and integration of the specific miRNA construct into differentiated and non-dividing cells, such as neurons and dendritic cells.

Autres remarques

Based on miRBase V20 Mature ID

Produits recommandés

Two negative controls are available: NCLMIR001 and NCLMIR002

Informations légales

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

RID ADR

UN 3245 9

WGK Allemagne

WGK 3

Flash Point(F)

Not applicable

Point d'éclair C

Not applicable

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique

Réseaux sociaux

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Recherche. Développement. Production

Nous sommes un des tout premiers fournisseurs du secteur mondial des Sciences de la vie proposant des solutions et des services pour la recherche, le développement et la production en biotechnologies, et le développement et la production de produits thérapeutiques pharmaceutiques.

© 2021 Merck KGaA, Darmstadt, Allemagne et/ou ses sociétés affiliées. Tous droits réservés.

La reproduction d'une quelconque partie du contenu de ce site est strictement interdite sans autorisation.