SRP6583

Sigma-Aldrich

EED (1-441) human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonyme(s):
Embryonic Ectoderm Development
NACRES:
NA.32

Source biologique

human

Recombinant

expressed in E. coli

Essai/Dosage

≥95% (SDS-PAGE)

Forme

liquid

Poids mol.

53.5 kDa (1- 441 aa + NT His Tag)

Conditionnement

pkg of 25 μg

N° d'accession UniProt

Expédié(e)(s) dans

dry ice

Temp. de stockage

−70°C

Gene Information

human ... EED(8726)

Description générale

Polycomb protein Embryonic Ectoderm Development (EED) is a WD repeat containing member of the Polycomb-group (PcG) family. The EED protein mediates repression of gene activity through histone deacetylation, and may act as a specific regulator of integrin function. EED is an established transcriptional repressor, as a novel NIPP1 interactor. NIPP1 only interacted with full-length EED, whereas two EED interaction domains were mapped to the central and COOH-terminal thirds of NIPP1. In mammalian cells, EED is present as four distinct isoforms, which are believed to be produced by utilizing four distinct, in-frame translation start sites in a common EED mRNA. The extra sex combs (esc) gene of Drosophila and its mammalian homologue embryonic ectoderm development (EED) play pivotal roles in establishing Polycomb-group (Pc-G) mediated transcriptional silencing of regulatory genes during early development. EED/Sox2 regulatory loop contributes to the maintenance of self-renewal in embryonic stem (ES) cells by controlling histone methylation and acetylation. EED has been defined by the inability of embryos homozygous for certain c deletions to develop beyond the early stages of gastrulation.

Forme physique

50 mM sodium phosphate, pH 7.2, containing 100 mM sodium chloride and 20% glycerol.

RID ADR

NONH for all modes of transport

WGK Allemagne

WGK 3

Flash Point(F)

Not applicable

Point d'éclair C

Not applicable

Certificat d'analyse
Certificat d'origine

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique

Réseaux sociaux

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Recherche. Développement. Production

Nous sommes un des tout premiers fournisseurs du secteur mondial des Sciences de la vie proposant des solutions et des services pour la recherche, le développement et la production en biotechnologies, et le développement et la production de produits thérapeutiques pharmaceutiques.

© 2021 Merck KGaA, Darmstadt, Allemagne et/ou ses sociétés affiliées. Tous droits réservés.

La reproduction d'une quelconque partie du contenu de ce site est strictement interdite sans autorisation.