Toutes les photos(1)

51101AST

Supelco

Astec® Cellulose DMP Chiral HPLC Column

5 μm particle size, L × I.D. 25 cm × 2.1 mm

eCl@ss:
32110501

Matériau

stainless steel column

Agence/méthode

suitable for USP L40

Gamme de produits

Astec®

Conditionnement

pkg of 1 ea

manufacturer/tradename

Astec®

Paramètre

0-40 °C temperature
50 bar pressure (725 psi)

Application(s)

HPLC: suitable

L × D.I.

25 cm × 2.1 mm

Matrice

fully porous particle
silica gel high purity, spherical
silica gel particle platform

Groupe de la matrice active

cellulose, DMPC- phase

Taille des particules

5 μm

Dimension de pores

100 Å pore size

operating pH

3.5-6.8

Technique de séparation

chiral

Vous recherchez des produits similaires ? Visit Guide de comparaison des produits

Catégories apparentées

Description générale

Astec® Cellulose DMP HPLC columns separate a wide range of chiral compounds under normal phase, polar organic, and SFC conditions. They exhibit high efficiency, high loading capacity, and excellent column lifetime. With performance comparable to other DMPC-derivatized cellulose CSPs, but at substantially lower price, Astec® Cellulose DMP is a must-have for every chiral column HPLC or SFC screening protocol. Astec® Cellulose DMP is complementary to the other Astec® CSPs, including CHIROBIOTIC®, CYCLOBOND® and the P-CAP product lines.

Description: Spherical, porous silica coated with DMPC (dimethylphenyl carbamate)-derivatized cellulose
Particle diameter: 5 μm
Field of Use: Small molecule enantiomer separations in normal phase, polar organic, and SFC modes

Produits recommandés

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Informations légales

Astec is a registered trademark of Sigma-Aldrich Co. LLC
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC
CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC
P-CAP is a trademark of Sigma-Aldrich Co. LLC

Certificat d'analyse

Certificat d'origine

Katrijn De Klerck et al.
Journal of pharmaceutical and biomedical analysis, 69, 77-92 (2012-03-01)
Chirality has a significant impact on drug discovery and development processes in the pharmaceutical industry. As the number of enantiopure drugs launched onto the market is yearly increasing, the need for fast and performant enantioseparation methods with minimal costs is
Katrijn De Klerck et al.
Journal of chromatography. A, 1328, 85-97 (2014-01-21)
Since their introduction on the market the applicability of immobilized polysaccharide-based chiral stationary phases in high-performance liquid chromatography has been thoroughly investigated. These immobilized phases have the benefit to be applicable with a wide range of modifiers, potentially extending the

Contenu apparenté

Astec Cellulose DMP

Separate a wide range of chiral compounds via normal phase, polar organic, and SFC conditions; useful in pharma, chemical, & environmental industry applications.

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique