Homocysteine causes vascular endothelial dysfunction by disrupting endoplasmic reticulum redox homeostasis.
Redox biology (2018-10-08)
Xun Wu, Lihui Zhang, Yütong Miao, Juan Yang, Xian Wang, Chih-Chen Wang, Juan Feng, Lei Wang
RÉSUMÉ
Endothelial dysfunction induced by hyperhomocysteinemia (HHcy) plays a critical role in vascular pathology. However, little is known about the role of endoplasmic reticulum (ER) redox homeostasis in HHcy-induced endothelial dysfunction. Here, we show that Hcy induces ER oxidoreductin-1α (Ero1α) expression with ER stress and inflammation in human umbilical vein endothelial cells and in the arteries of HHcy mice. Hcy upregulates Ero1α expression by promoting binding of hypoxia-inducible factor 1α to the ERO1A promoter. Notably, Hcy rather than other thiol agents markedly increases the GSH/GSSG ratio in the ER, therefore allosterically activating Ero1α to produce H2O2 and trigger ER oxidative stress. By contrast, the antioxidant pathway mediated by ER glutathione peroxidase 7 (GPx7) is downregulated in HHcy mice. Ero1α knockdown and GPx7 overexpression protect the endothelium from HHcy-induced ER oxidative stress and inflammation. Our work suggests that targeting ER redox homeostasis could be used as an intervention for HHcy-related vascular diseases.
MATÉRIAUX
Référence de produit
Marque
Description du produit
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
Monoclonal Anti-β-Actin−Peroxidase antibody produced in mouse, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Diamide
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
NG-Methyl-L-arginine acetate salt, ≥98% (TLC)
Sigma-Aldrich
Monoclonal Anti-GAPDH−Peroxidase antibody produced in mouse, clone GAPDH-71.1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Biphenylene, 99%
Sigma-Aldrich
Anti-Sulfenic Acid Modified Cysteine (2-Thiodimedone-Specific Ig) Antibody, serum, from rabbit
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, Vitamin B12 deficient
Sigma-Aldrich
Anti-GRP78/BiP (GL-19) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Ero1 Antibody, clone 2G4/12, clone 2G4/12, from mouse
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human HIF1A
Réseaux sociaux
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Recherche. Développement. Production
Nous sommes un des tout premiers fournisseurs du secteur mondial des Sciences de la vie proposant des solutions et des services pour la recherche, le développement et la production en biotechnologies, et le développement et la production de produits thérapeutiques pharmaceutiques.
© 2021 Merck KGaA, Darmstadt, Allemagne et/ou ses sociétés affiliées. Tous droits réservés.
La reproduction d'une quelconque partie du contenu de ce site est strictement interdite sans autorisation.