Homeostatic regulation of STING protein at the resting state by stabilizer TOLLIP.
Nature immunology (2020-01-15)
Vladislav Pokatayev, Kun Yang, Xintao Tu, Nicole Dobbs, Jianjun Wu, Robert G Kalb, Nan Yan
RÉSUMÉ
STING (stimulator of interferon genes) is an important innate immune protein, but its homeostatic regulation at the resting state is unknown. Here, we identified TOLLIP as a stabilizer of STING through direct interaction to prevent its degradation. Tollip deficiency results in reduced STING protein in nonhematopoietic cells and tissues, and renders STING protein unstable in immune cells, leading to severely dampened STING signaling capacity. The competing degradation mechanism of resting-state STING requires IRE1α and lysosomes. TOLLIP mediates clearance of Huntington's disease-linked polyQ protein aggregates. Ectopically expressed polyQ proteins in vitro or endogenous polyQ proteins in Huntington's disease mouse striatum sequester TOLLIP away from STING, leading to reduced STING protein and dampened immune signaling. Tollip-/- also ameliorates STING-mediated autoimmune disease in Trex1-/- mice. Together, our findings reveal that resting-state STING protein level is strictly regulated by a constant tug-of-war between 'stabilizer' TOLLIP and 'degrader' IRE1α-lysosome that together maintain tissue immune homeostasis.
MATÉRIAUX
Référence de produit
Marque
Description du produit
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
DMXAA, ≥98% (HPLC), solid
Sigma-Aldrich
4μ8C, ≥98% (HPLC)
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule) antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Réseaux sociaux
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Recherche. Développement. Production
Nous sommes un des tout premiers fournisseurs du secteur mondial des Sciences de la vie proposant des solutions et des services pour la recherche, le développement et la production en biotechnologies, et le développement et la production de produits thérapeutiques pharmaceutiques.
© 2021 Merck KGaA, Darmstadt, Allemagne et/ou ses sociétés affiliées. Tous droits réservés.
La reproduction d'une quelconque partie du contenu de ce site est strictement interdite sans autorisation.