BR471500

BRAND® staining trough, black

POM

Synonym(s):
Staining, Staining Accessories, Microscopy
NACRES:
MA.01

material

POM

description

For use with black staining tray

manufacturer/tradename

BRAND 471500

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service