Merck
All Photos(1)

1.15101

Millipore

Silica gel 60 (0.063-0.100 mm)

for column chromatography

Synonym(s):
Silica gel
CAS Number:
MDL number:
EC Index Number:
231-545-4

material

spherical silica gel

Quality Level

particle size

63-200 μm

pore size

6 nm mean pore size

pH

7 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

bulk density

200‑1430 kg/m3

application(s)

plasma purification

separation technique

reversed phase

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

Inchi Key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

for column chromatography

Application

Silica gel 60 (0.063-0.100 mm) for column chromatography. CAS No. 7631-86-9, EC Number 231-545-4.

Analysis Note

Appearanceamorphous, fine powderColorwhitepH-value (10 % suspension)6.5 - 7.5Fe (Iron)≤ 0.02 %Loss on drying (150 °C)≤ 7 %Particle sizeminimum 90 % between 0.063 - 0.100 mm

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service