Merck
  • 124032
All Photos(1)

124032

Sigma-Aldrich

Akt Inhibitor XIV

The Akt Inhibitor XIV, also referenced under CAS 1191951-57-1, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):
Akt Inhibitor XIV, 4-Dodecyl-N-(1,3,4-thiadiazol-2-yl)benzenesulfonamide, PDK1 Inhibitor III, PHT-427
Empirical Formula (Hill Notation):
C20H31N3O2S2
CAS Number:
Molecular Weight:
409.61

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C20H31N3O2S2/c1-2-3-4-5-6-7-8-9-10-11-12-18-13-15-19(16-14-18)27(24,25)23-20-22-21-17-26-20/h13-17H,2-12H2,1H3,(H,22,23)

InChI key

BYWWNRBKPCPJMG-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

1 of 4

This Item
124011124020124037
Akt Inhibitor XIV The Akt Inhibitor XIV, also referenced under CAS 1191951-57-1, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Sigma-Aldrich

124032

Akt Inhibitor XIV

Akt Inhibitor IV The Akt Inhibitor IV, also referenced under CAS 681281-88-9, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Sigma-Aldrich

124011

Akt Inhibitor IV

Akt Inhibitor X The Akt Inhibitor X, also referenced under CAS 925681-41-0, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Sigma-Aldrich

124020

Akt Inhibitor X

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

form

solid

form

solid

form

solid

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

storage condition

OK to freeze, desiccated, protect from light

color

white

color

yellow

color

white

color

pink

solubility

DMSO: 50 mg/mL

solubility

DMSO: 5 mg/mL

solubility

water: 1 mg/mL

solubility

DMSO: 100 mg/mL

General description

A cell-permeable thiadiazolyl compound that binds to PH (pleckstrin homology) domain (Kd = 40.8 µM, Ki = 2.4 µM) and acts as a competitive and reversible blocker of Akt translocation to the cellular membrane with selectivity over PDK1 (Kd = 90.1 µM, Ki = 5.5 µM). Inhibits pAkt-Ser473 (IC50 = 6.3, 8.6 and 10 µM in Panc-1, BxPC-3 and MiaPaCa-2 cells, respectively), and induces apoptosis at 20 µM (40% and 31%) and causes growth arrest (IC50 = 65 and 30 µM) in Panc-1 and MiaPaCa-2 cells, respectively. Further, suppresses tumor growth and regression in mouse xenograft model (125 mg/kg, i.p.) and reduces pGSK-3α-Ser21 and pGSK-3β-Ser9 and p70S6K-Thr389 levels.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Meuillet, E.J., et al. 2010. Mol. Cancer Ther. 9, 706.
Moses, S.A., et al. 2009. Cancer Res. 69, 5073.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service