Skip to Content
Merck

124034

Akt Inhibitor XV, Isozyme-Selective

The Akt Inhibitor XV, Isozyme-Selective controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Akt Inhibitor XV, Isozyme-Selective, 5-Hydroxy-3-phenyl-2-(4-((4-(5-pyridin-2-yl-1H-1,2,4-triazol-3-yl)piperdin-1-yl)methyl)phenyl)-1,6-naphthyridine

Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C33H29N7O
Molecular Weight:
539.63
UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable naphthyridinone compound that acts as an allosteric Akt1/2-selective inhibitor (IC50 = 3.5, 42, and 1900 nM against Akt1, Akt2, and Akt3, respectively) in a PH domain-dependent, but not ATP-competitive, manner, with little activity against a panel of more than 160 other kinases, including the related AGC family kinases (inactive at 10 µM against GSK, PDK1, PKA, PKC, and SGK). Effectively inhibits the basal Akt phosphorylation in cancer cells with deregulated Akt pathway activation, including the HCT116 colorectal carcinoma and a panel of breast cancer cell lines (IC50<500 nM), and suppresses Akt-dependent cell growth both in cultures in vitro (IC50 ≤1 µM) and in mice in vivo (50 to 100 mg/kg, SC). Although also referred to as AKTi-1/2, this is not the same compound as Akt Inhibitor XV (Cat. Nos. 124017 & 124018).

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

She, Q.B., et al. 2010. Cancer Cell18, 39.
Bilodeau, M.T., et al. 2008. Bioorg. Med. Chem. Lett.18, 3178.
She, Q.B, et al. 2008. PLoS One3, e3065.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service