Merck
All Photos(2)

Documents

124037

Sigma-Aldrich

Akt Inhibitor XIX, 3CAI

The Akt Inhibitor XIX, 3CAI, also referenced under CAS 28755-03-5, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):
Akt Inhibitor XIX, 3CAI, 3-Chloroacetyl-indole, 2-Chloro-1-(1H-indol-3-yl)ethanone
Empirical Formula (Hill Notation):
C10H8ClNO
CAS Number:
Molecular Weight:
193.63
MDL number:

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

pink

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C10H8ClNO/c11-5-10(13)8-6-12-9-4-2-1-3-7(8)9/h1-4,6,12H,5H2

InChI key

LLZQFAXTCYDVTR-UHFFFAOYSA-N

General description

A cell-permeable chloroacetyl-indole compound that inhibits Akt1 and Akt2 kinase activity (IC50<1 µM) by directly targeting Akt PH domain, exhibiting little or much reduced activity against a panel of 84 other kinases, including PI-3 Kinase (60% inhibition at 10 µM). Effectively inhibits Akt-mediated cellular GSK3β Ser9 and mTOR Ser2448 phosphorylation (4 µM in HCT116 cultures) in a time-dependent manner, resulting in effective apoptosis induction in HCT116 and HT29 cultures (by 55% and 60%, respectively; 4 µM for 4 days). Reported to retard HCT116-derived tumor growth by 50% when administered at an oral dosage of 30 mg/kg in mice (5 times/wk for 3 wks) in vivo.
A cell-permeable chloroacetyl-indole compound that inhibits Akt1 and Akt2 kinase activity (IC50<1 µM) by directly targeting Akt PH domain, exhibiting little or much reduced activity against a panel of 84 other kinases. Effectively inhibits Akt-mediated downstream effector proteins phosphorylation in a time-dependent manner, resulting in effective apoptosis induction in HCT116 and HT29 cultures (by 55% and 60%, respectively; 4 µM for 4 days). Reported to retard HCT116-derived tumor growth in mice (EC50 = 30 mg/kg; 5 times/wk for 3 wks via p.o.) in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kim, D.J., et al. 2011. Cancer Prev. Res.4, 1842.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service