Skip to Content
Merck

178612

Anti-Aquaporin 2 (254-271) Rabbit pAb

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
51111800
Species reactivity:
(human, mouse, rat)
Species reactivity:
-
Application:
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

form

lyophilized

species reactivity

(human, mouse, rat)

storage condition

OK to freeze

dilution

(Immunoblotting (1:200-1:600)
Immunohistochemistry
Immunoprecipitation )

isotype

IgG

storage temp.

−20°C

General description

Protein A purified rabbit polyclonal antibody. Recognizes the ~40 kDa and ~29 kDa aquaporin-2 proteins. Supplied with a control peptide (AQP2254-271 immunogen peptide).

Immunogen

Mouse/Rat

Application

Immunoblotting (1:200-1:600)Immunohistochemistry Immunoprecipitation


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library