Skip to Content
Merck

219350

Anti-β-Catenin, Dephospho Mouse mAb (8E4)

lyophilized, clone 8E4, Calbiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352203
Clone:
8E4, monoclonal
Species reactivity:
human, rat, mouse
Application:
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8E4, monoclonal

form

lyophilized

species reactivity

human, rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

General description

Mouse monoclonal antibody purified by thiophilic adsorption and size exclusion chromatography. Recognizes the ~90 kDa nuclear dephospho β-catenin protein. Supplied with a control lysate derived from 2x106 LiCl-treated SW480 cells.
Recognizes the ~90 kDa nuclear dephospho-β-catenin in vanadate treated A431 cells. Supplied with a control lysate derived from 2x106 LiCl-treated SW480 cells.
This Anti-β-Catenin, Dephospho Mouse mAb (8E4) is validated for use in ELISA, Immunoblotting, Immunoprecipitation for the detection of β-Catenin, Dephospho.

Immunogen

Mouse
a synthetic peptide corresponding to amino acids 27-37 of mouse β-catenin, conjugated to KLH

Application

ELISA (0.05 µg/ml)

Immunoblotting (1 µg/ml colorimetric, 0.1 µg/ml, chemiluminescence)

Immunoprecipitation (1-10 µg/10⁶ LiCl-treated SW480 cells)

Physical form

100 µg antibody lyophilized from 2X PBS, sucrose, PEG and 200 µl lyophilized control lysate.

Preparation Note

Reconstitute antibody with 1 ml dH₂O. Following reconstitution, refrigerate (4°C) for short-term storage or aliqout, flash-freeze in liquid nitrogen, and deep freeze (-70°C) for long-term storage. Stock solutions are stable for up to 3 months at 4°C or for up to 1 year at -70°C. Thaw aliquots at 37°C. Reconstitute positive control lysate with 200 µl H₂O. After complete solubilization of the proteins add 200 µl SDS-PAGE sample buffer and incubate at 90°C for 5 min. Following reconstitution of positive control, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions.

Analysis Note

Positive Control
LiCl-treated SW480 cells (included)

Other Notes

SW480 LiCl cell lysate is recommended for immunoblot applications at a concentration of 20 µl/lane (mini gel) for chemiluminescent detection. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
219350-1SET04055977202168

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service