All Photos(1)

MCEP12H48

Millipore

Millicell® Hanging Cell Culture Insert, PET 8 μm, 12-well, 48/pk

Polyethylene Terephthalate hanging cell culture insert with pore size of 8.0 μm used in a 12-well plate for cell attachment, cell culture, cell differentiation & ICC.

Synonym(s):
tissue culture plate, tissue culture insert, permeable culture insert, cell culture inserts

Quality Level

material

PET membrane
flat wells
polystyrene housing

sterility

sterile

feature

lid: no

packaging

pack of 48 ea (individually blister packaged)

parameter

50 °C max. temp.

filter diam.

12 mm

filtration area

1.1 cm2

size

12 wells

well maximum volume

1.4 mL

working volume

400 μL

pore size

8.0 μm

input

sample type epithelial cells
sample type induced pluripotent stem cell(s)
sample type cardiac stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type hematopoietic stem cell(s)
sample type mesenchymal stem cell(s)
sample type pancreatic stem cell(s)
sample type: human embryonic stem cell(s)
sample type neural stem cell(s)

surface area

1.1 cm2

binding type

Tissue Culture (TC)-treated surface

General description

Polyethylene Terephthalate (PET) hanging cell culture insert with pore size of 8.0 μm used in a 12-well plate for cell attachment, cell culture, cell differentiation & ICC.Device configuration: 12-well plate, 8μm pore sizeMillicell® cell culture inserts are an easy to use and convenient device for adherent or suspension cells. Allowing the access of media from both their apical and basolateral sides. Cell growth, structure, and function more closely mimic what occurs in vivo. In addition, Millicell® inserts make it possible to study both sides of the cell monolayer.
Millicell® Hanging Inserts: Preloaded in receivers or available standalone
  • For co-culturing and permeability assays
  • Unique design allows easier basolateral access than other hanging inserts with less risk of contamination
  • Available in 5 pore sizes and 3 diameters, including a 1 μm pore size that is optically transparent for better visualization by microscopy

Applications: Cell Attachment, Cell Growth, Cell Differentiation, Immunocytochemistry

Application

Polyethylene Terephthalate hanging cell culture insert with pore size of 8.0 μm used in a 12-well plate for cell attachment, cell culture, cell differentiation & ICC.

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificate of Analysis

Certificate of Origin

Protocols

Microscopic Examination of Samples

This protocol covers 3 modes for the microscopic examination of cell samples.

Indirect Co-culture of Embryonic Stem Cells with Embryonic Fibroblasts

This page covers the indirect co-culture of embryonic stem cells with embryonic fibroblasts.

Related Content

ECM Coating Protocols for Millicell®

This page covers the ECM coating protocols developed for four types of ECMs on Millicell®-CM inserts, Collagen Type 1, Fibronectin, Laminin, and Matrigel.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service