All Photos(1)
CLS3081
Corning® 96 well storage mat applicator
non-sterile
NACRES:
NB.15
sterility
non-sterile
packaging
case of 1 ea
pack of 1 ea
General description
  • Corning® 96 well storage mat applicator aids in evenly applying storage mats to Corning® 96 well polypropylene plates, blocks, and 96 well polystyrene plates.
  • Sealing pressure to be adjusted using setting gage.
  • Two aluminum riser blocks enable sealing plates of various sizes.
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificate of Analysis
Certificate of Origin