Merck
All Photos(1)

Documents

SAB4701067

Sigma-Aldrich

Monoclonal Anti-CD157-APC antibody produced in mouse

clone SY11B5

Sign Into View Organizational & Contract Pricing

Synonym(s):
BST1

biological source

mouse

conjugate

Allophycocyanin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SY11B5, monoclonal

form

buffered aqueous solution

technique(s)

flow cytometry: 10 μL

accession no.

BC012095

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... BST1(683)

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB4700023SAB4701134SAB4701071
Gene Information

human ... BST1(683)

Gene Information

human ... BSG(682)

Gene Information

human ... CD160(11126)

Gene Information

-

conjugate

Allophycocyanin conjugate

conjugate

Allophycocyanin conjugate

conjugate

Allophycocyanin conjugate

conjugate

Allophycocyanin conjugate

clone

SY11B5, monoclonal

clone

MEM-M6/1, monoclonal

clone

BY55, monoclonal

clone

TB01, monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

technique(s)

flow cytometry: 10 μL

technique(s)

flow cytometry: suitable

technique(s)

flow cytometry: 10 μL

technique(s)

flow cytometry: 10 μL

Still not finding the right product?  

Give our Product Selector Tool a try.

General description

The mouse monoclonal antibody SY11B5 recognizes CD157, an approximately 45 kDa GPI-anchored protein expressed mainly on monocytes, macrophages, granulocytes and bone marrow stromal cells.

Immunogen

Human CD157

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service