Merck
All Photos(3)

Documents

WH0010225M1

Sigma-Aldrich

Monoclonal Anti-CD96 antibody produced in mouse

clone 3H8, purified immunoglobulin, buffered aqueous solution

Synonym(s):
Anti-CD96 antigen, Anti-DKFZp667E2122, Anti-MGC22596, Anti-TACTILE
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3H8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank® accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CD96(10225)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB1410073SAB1404801SAB1406950
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

clone

3H8, monoclonal

clone

polyclonal

clone

1C8, monoclonal

clone

polyclonal

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

General description

CD96 (cluster of differentiation 96) belongs to the IgG (immunoglobulin) superfamily receptors. It is expressed in the spinal cord, kidney, trachea, digestive tissues, prostate, placenta, bone of adults and fetal brain and liver. This gene is located on human chromosome 3q13.
The protein encoded by this gene belongs to the immunoglobulin superfamily. It is a type I membrane protein. The protein may play a role in the adhesive interactions of activated T and NK cells during the late phase of the immune response. It may also function in antigen presentation. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. (provided by RefSeq)

Immunogen

CD96 (AAH20749, 1 a.a. ~ 402 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEKKWKYCAVYYIIQIHFVKGVWEKTVNTEENVYATLGSDVNLTCQTQTVGFFVQMQWSKVTNKIDLIAVYHPQYGFYCAYGRPCESLVTFTETPENGSRWTLHLRNMSCSVSGRYECMLVLYPEGIQTKIYNLLIQTHVTADEWNSNHTIEIEINQTLEIPCFQNSSSKISSEFTYAWSVEDNGTQETLISQNHLISNSTLLKDRVKLGTDYRLHLSPVQIFDDGRKFSCHIRVGPNKILRSSTTVKVFAKPEIPVIVENNSTDVLVERRFTCLLKNVFPKANITWFIDGSFLHDEKEGIYITNEERKGKDGFLELKSVLTRVHSNKPAQSDNLTIWCMALSPVPGNKVWNISSGKITFLLGSEISSTDPPLSVTESTLDTQPSPASSVSPASKNVFTLSY

Biochem/physiol Actions

CD96 (cluster of differentiation 96) plays major inhibitory roles in immune function. It helps to increase the cytotoxicity of natural killer (NK) cells. Mutations in CD9 result in C (Opitz Trigonocephaly) Syndrome.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Mutations in CD96, a member of the immunoglobulin superfamily, cause a form of the C (Opitz trigonocephaly) syndrome
Kaname T, et al.
American Journal of Human Genetics, 81(4), 835-841 (2007)
Differential expression of CD96 surface molecule represents CD8? T cells with dissimilar effector function during HIV-1 infection
Eriksson EM, et al.
PLoS ONE (2012)
Molecular Pathways: Targeting CD96 and TIGIT for Cancer Immunotherapy
Blake SJ, et al.
Clinical Cancer Research, 22(21), 5183-5188 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service