Z743320

Bake Tags oven-safe labels

L × W 1.50 in. × 0.75 in., white, roll of 250 ×

NACRES:
NB.35

manufacturer/tradename

Diversified Biotech BAKE-250

packaging

roll of 250 ×

application(s)

PCR: suitable

L × W

1.50 in. × 0.75 in.

color

white

General description

Bake Tags are oven-safe marking labels that withstand conventional oven-sterilization (typically 180°C) or heating to inactivate RNase, DNase and other enzymes. Bake Tags will withstand autoclaving, cryogenic freezer storage conditions and boiling baths without peeling or falling off.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service