Skip to Content
Merck
Sign Into View Organizational & Contract Pricing

About This Item

CAS Number:
Molecular Weight:
785.03
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

2-((2-((3,12-dihydroxy-17-(2-hydroxy-6-methyloct-5-en-2-yl)-4,4,10,14-tetramethylhexadecahydro-1H-cyclopenta[a]phenanthren-6-yl)oxy)-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-methyltetrahydro-2H-pyran-3,4,5-triol

Inchi Code

1S/C42H72O13/c1-9-20(2)11-10-14-42(8,51)22-12-15-40(6)24-18-26(36-39(4,5)28(45)13-16-41(36,7)23(24)17-25(44)29(22)40)53-38-35(33(49)31(47)27(19-43)54-38)55-37-34(50)32(48)30(46)21(3)52-37/h11,21-38,43-51H,9-10,12-19H2,1-8H3

InChI key

QOVKKEKBURQILF-UHFFFAOYSA-N

storage temp.

4

Purity

99.50%

Country of Origin

CN

Shipping Temperature

Room temperature/blue ice

Physical Form

solid

Product Link

https://www.targetmol.com/compound/Ginsenoside Rg2

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service