Merck
Cell Therapy Manufacturing Systems
The ekko™ Acoustic Cell Processing System is a breakthrough technology that provides freedom from process limitations in cell therapy manufacturing. Its unique acoustic technology provides a gentle way of processing cells while maintaining cell quality. Its unmatched ease-of-use and flexibility support multiple cell applications, process steps, and scales.
The ekko™ System is flexible for a range of cell types and variations including without limitation to:
  • Wash
  • Concentration
  • Media Exchange
  • Low Volume Processing
  • Formulation
Products
Key Benefits
  • High cell recovery and viability with residual removal of >95%
  • Simple and ergonomic installation of ekko™ Single-Use Assemblies
  • ekko™ Software and ekko™ Architect, our flexible protocol development tool, is designed for easy navigation and use
  • Freedom from process limitations - for use in multiple unit operations across a variety of cell types
  • Low volume processing enabling collection of 5 mL cell concentrate
  • 21 CFR Part 11 compliance-ready and developed under GAMP5 guidelines
The ekko™ Acoustic Cell Processing System is flexible for a range of cell types and variations including without limitation to:
  • T cells
  • Pluripotent Stem Cells (iPSCs)
  • Mesenchymal Stem Cells (MSCs)
  • Hematopoietic Stem Cells (HSCs)
Concentrate display screen example for ekko Software
ekko™ Software and ekko™ Architect
Intuitive Navigation
Our touchscreen interface allows for fast navigation and quick response rates that support increased productivity.
Step-by-Step Instructions
Graphical step-by-step instructions reduce error and simplify training and technology transfer.
Multiple Modes of Operation
The ekko™ Software offers two (2) modes of operation:
  • The Development Mode allows for process development and protocol optimization.
  • The Manufacturing Mode locks procedures for execution in compliance with 21 CFR Part 11.
ekko System Single-Use Assembly
ekko™ Single use Assembly
Ready to Process in Minutes
Our single-use assembly installs in less than a minute. Simple tube welds connect incoming material, product, and waste bags.
Error Proof Set-up
We have designed our single-use assembly for error-proof installation. Latches ensure secure attachment before starting your run. In addition, to improve operational safety and ease of use, all weldable tubes are labeled for easy identification.
Ergonomic Design
We have ergonomically designed the single-use assembly to integrate all fluid path components into a cartridge format that circumvents the need for manual tube threading through pinch valves or incorrect orientation of lines. In addition, the ekko™ Software controls all functions to ensure reliable and repeatable operation.
Maximum Operational Flexibility
We provide two tubing options for each weldable tube line: Thermoplastic elastamor (TPE) and Polyvinyl chloride (PVC). These options maximize operational flexibility, allowing you to select the correct transfer assemblies for your needs.