Skip to Content
Merck
Anti-APOB antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-APOB antibody produced in rabbit

Anti-APOB antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Products

Product Category
Brand
Biological Source
Clonality
Physical Form
Product Line
Research Area
Conjugate
Quality Segment
Species Reactivity
Technique