
Anti-OVOL1 antibody produced in rabbit

Synonym(s):
OVOL1, Putative transcription factor Ovo-like 1, hOvo1
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique