Skip to Content
GB
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methyldec-9-enoic acid
CAS Number:
288617-75-4
Molecular Weight:
421.54
Products
Brand
BLD Pharmatech (1)
Fluorochem (Preferred Partner) (1)
Synthonix Corporation (1)
Purity
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
FLUH99C8FF6C
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methyldec-9-enoic acid,
Expand
View Pricing
SY3H6E4146EC
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methyldec-9-enoic acid,
Expand
View Pricing
BL3H1F1C7DB4
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methyldec-9-enoic acid,
Expand
View Pricing