Skip to Content
Merck
C646 ≥98% (HPLC)

C646

C646 ≥98% (HPLC)
Synonym(s):
Histone Acetyltransferase p300 Inhibitor, C646, 4-[4-[[5-(4,5-Dimethyl-2-nitrophenyl)-2-furanyl]methylene]-4,5-dihydro-3-methyl-5-oxo-1H-pyrazol-1-yl]benzoic acid
Empirical Formula (Hill Notation):
C24H19N3O6
CAS Number:
Molecular Weight:
445.42

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-2 of 2