Skip to Content
Merck
Millex™ polyethersulfone syringe filter pore size 0.22 μm, diam. 25 mm, non-sterile, hydrophilic

Millex polyethersulfone syringe filter

Millex™ polyethersulfone syringe filter pore size 0.22 μm, diam. 25 mm, non-sterile, hydrophilic

Products

Product Category
Brand
Color
Manufacturer
Product Line
Agency
Diameter (mm)
Height (mm)
Surface Area cm²
Volume (ml)
Quality Segment
Hydrophobicity
Application
Housing Material
Membrane Material
Package Size
Pore size (µm)
Sterility
Sterilization Method
Technique
Showing 1-11 of 11