Skip to Content
Merck
Millex™ PVDF syringe filter pore size 0.45 μm, diam. 25 mm, non-sterile, hydrophilic

Millex PVDF syringe filter

Millex™ PVDF syringe filter pore size 0.45 μm, diam. 25 mm, non-sterile, hydrophilic

Products

Product Category
Brand
Color
Product Line
Diameter (mm)
Height (mm)
Surface Area cm²
Volume (ml)
Quality Segment
Hydrophobicity
Housing Material
Membrane Material
Package Size
Pore size (µm)
Sterility
Sterilization Method
Technique
Showing 1-20 of 25
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2