GB
EN
Applications
Products
Services
Support
Home
Search Results
Sulfuric acid 25%
(1)
Sulfuric acid 25%
Synonyms:
Sulfuric acid solution
Linear Formula:
H
2
SO
4
CAS Number:
7664-93-9
Molecular Weight:
98.08
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
1.00716
Sulfuric acid 25%, for analysis EMSURE
®
Expand
View Pricing