Merck
Sulfuric acid 95-97% for analysis EMPARTA® ACS

Sulfuric acid 95-97%

Sulfuric acid 95-97% for analysis EMPARTA® ACS
Synonyms:
Sulfuric acid
Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Sulfuric acid 95-97%, for analysis EMSURE® ISO
Sulfuric acid 95-97%, for analysis (max. 0.005 ppm Hg) EMSURE® ACS,ISO,Reag. Ph Eur
Sulfuric acid 95-97%, for analysis EMPARTA® ACS