Merck
Sulfuric acid 96% for the determination of viscosity acc. to DIN EN ISO 307

Sulfuric acid 96%

Sulfuric acid 96% for the determination of viscosity acc. to DIN EN ISO 307
Synonyms:
Sulfuric acid
Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Sulfuric acid 96%, Suprapur®
Sulfuric acid 96%, Ultrapur
Sulfuric acid 96%, for the determination of viscosity acc. to DIN EN ISO 307