Merck
  • Discovery of 4-amino and 4-hydroxy-1-aroylindoles as potent tubulin polymerization inhibitors.
Discovery of 4-amino and 4-hydroxy-1-aroylindoles as potent tubulin polymerization inhibitors.
Journal of medicinal chemistry (2008-07-01)
Jing-Ping Liou, Zi-Yi Wu, Ching-Chuan Kuo, Chi-Yen Chang, Pei-Yi Lu, Chi-Ming Chen, Hsing-Pang Hsieh, Jang-Yang Chang
ABSTRACT
1-Aroylindoline, 1-aroyl-1,2,3,4-tetrahydroquinoline, and 1-aroylindole derivatives were synthesized and evaluated for anticancer activity. The 4-amino and 4-hydroxy-1-aroylindoles 26 and 27 with IC 50 of 0.9 and 0.6 microM, respectively, exhibited antitubulin activity superior or comparable to that of colchicine and combretastatin A-4. They also showed antiproliferative activity with IC 50 of 0.3-5.4 nM in a set of human cancer cell lines.
Sigma-Aldrich
Colchicine, suitable for plant cell culture, BioReagent, ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Pricing and availability is not currently available.